Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PBXIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075265.1 | NP_001068733.1 | 9 - 10 | 931 | 64 | |
GenBank mRNA | BC148133.1 | - | 9 - 10 | 986 | 64 | |
BT026164.1 | - | 9 - 10 | 931 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |