Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRR19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075635.1 | NP_001069103.1 | 3 - 4 | 1177 | 98 | |
GenBank mRNA | BC119867.1 | - | 3 - 4 | 1177 | 98 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Bos taurus Probe spans exons |