Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBM23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075636.1 | NP_001069104.1 | 3 - 4 | 404 | 84 | |
GenBank mRNA | BC122593.1 | - | 3 - 4 | 404 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |