Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM81B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075688.2 | NP_001069156.1 | 6 - 7 | 795 | 123 | |
GenBank mRNA | BC122751.1 | - | 6 - 7 | 796 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |