Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNAP29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075959.1 | NP_001069427.1 | 1 - 2 | 318 | 86 | |
GenBank mRNA | BC122755.1 | - | 1 - 2 | 318 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |