Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAAF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075990.1 | NP_001069458.1 | 2 - 3 | 101 | 56 | |
GenBank mRNA | BC118307.1 | - | 2 - 3 | 101 | 56 | |
BT025484.1 | - | 2 - 3 | 84 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |