Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARRDC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076257.2 | NP_001069725.1 | 6 - 7 | 1268 | 68 | |
GenBank mRNA | BC120276.1 | - | 6 - 7 | 1269 | 68 | |
KP292570.1 | - | 6 - 7 | 1269 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |