Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BPIFA2C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076503.1 | NP_001069971.1 | 7 - 8 | 916 | 116 | |
GenBank mRNA | DQ835286.1 | - | 7 - 8 | 916 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Bos taurus Probe spans exons |