Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DZANK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076838.1 | NP_001070306.1 | 14 - 15 | 1549 | 97 | |
GenBank mRNA | BC123405.1 | - | 14 - 15 | 1549 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |