Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076849.1 | NP_001070317.1 | 2 - 3 | 299 | 90 | |
GenBank mRNA | BC123847.1 | - | 2 - 3 | 299 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |