Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CACNG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078014.1 | NP_001071482.1 | 1 - 2 | 636 | 133 | |
GenBank mRNA | BC119905.1 | - | 1 - 2 | 636 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |