Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: METTL9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080367.2 | NP_001073836.1 | 3 - 4 | 716 | 70 | |
GenBank mRNA | BC120133.1 | - | 3 - 4 | 717 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |