Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLRB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174071.2 | NP_776496.1 | 4 - 5 | 367 | 122 | |
GenBank mRNA | AF268376.1 | - | 3 - 4 | 410 | 122 | |
BC123491.1 | - | 4 - 5 | 367 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |