Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGS16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174450.3 | NP_776875.1 | 1 - 2 | 140 | 84 | |
GenBank mRNA | BC123439.1 | - | 1 - 2 | 140 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |