Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NFE2L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011678.2 | NP_001011678.2 | 3 - 4 | 491 | 78 | |
GenBank mRNA | AB162435.1 | - | 3 - 4 | 400 | 78 | |
BC102275.1 | - | 3 - 4 | 540 | 78 | ||
BT030494.1 | - | 3 - 4 | 491 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |