Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077020.1 | NP_001070488.1 | 6 - 7 | 579 | 88 | |
GenBank mRNA | BC105572.1 | - | 6 - 7 | 595 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |