Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NHEDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077892.2 | NP_001071360.1 | 3 - 4 | 316 | 72 | |
GenBank mRNA | BC126670.1 | - | 3 - 4 | 317 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |