Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GSDMA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078090.1 | NP_001071558.1 | 5 - 6 | 717 | 75 | |
GenBank mRNA | BC126753.1 | - | 5 - 6 | 717 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |