Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDIT4L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081519.1 | NP_001074988.1 | 2 - 3 | 298 | 81 | |
GenBank mRNA | BC133398.1 | - | 2 - 3 | 298 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |