Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HPS6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081586.1 | NP_001075055.1 | 1 | 2447 | 77 | |
GenBank mRNA | BC133397.1 | - | 1 | 2447 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Bos taurus Both primers and probe map within a single exon |