Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082434.2 | NP_001075903.1 | 4 - 5 | 862 | 73 | |
GenBank mRNA | BC133569.1 | - | 4 - 5 | 863 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |