Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BRMS1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083428.1 | NP_001076897.1 | 9 - 10 | 869 | 111 | |
GenBank mRNA | BC123661.1 | - | 9 - 10 | 869 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |