Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CA8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083690.2 | NP_001077159.1 | 5 - 6 | 868 | 103 | |
GenBank mRNA | BC134783.1 | - | 5 - 6 | 869 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |