Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC35F6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083704.1 | NP_001077173.1 | 4 - 5 | 607 | 62 | |
GenBank mRNA | BC134546.1 | - | 4 - 5 | 607 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |