Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM255A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083721.1 | NP_001077190.1 | 8 - 9 | 845 | 93 | |
GenBank mRNA | BC134568.1 | - | 8 - 9 | 845 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |