Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTC9C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083792.1 | NP_001077261.1 | 2 - 3 | 778 | 69 | |
GenBank mRNA | BC134555.1 | - | 2 - 3 | 778 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Bos taurus Probe spans exons |