Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WNT2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099363.2 | NP_001092833.1 | 2 - 3 | 687 | 62 | |
GenBank mRNA | BC146036.1 | - | 2 - 3 | 688 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |