Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD79A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174266.4 | NP_776691.2 | 2 - 3 | 395 | 90 | |
GenBank mRNA | BC120260.1 | - | 2 - 3 | 396 | 90 | |
D16412.1 | - | 2 - 3 | 370 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |