Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FNTA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177498.3 | NP_803464.2 | 7 - 8 | 847 | 116 | |
GenBank mRNA | M74083.1 | - | 7 - 8 | 731 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Bos taurus Probe spans exons |