Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GIN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024473.2 | NP_001019644.2 | 1 - 2 | 33 | 123 | |
GenBank mRNA | BC114679.1 | - | 1 - 2 | 33 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |