Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ELF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098909.2 | NP_001092379.1 | 4 - 5 | 608 | 103 | |
GenBank mRNA | BC142205.1 | - | 3 - 4 | 877 | 103 | |
EE353432.1 | - | 4 - 5 | 608 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |