Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TEX26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098913.2 | NP_001092383.1 | 5 - 6 | 662 | 61 | |
GenBank mRNA | BC146109.1 | - | 5 - 6 | 663 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |