Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDK20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098933.1 | NP_001092403.1 | 7 - 8 | 1104 | 60 | |
GenBank mRNA | BC146216.1 | - | 7 - 8 | 1104 | 60 | |
BC149150.1 | - | 1 - 2 | 300 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |