Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC80 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098982.2 | NP_001092452.1 | 3 - 4 | 2770 | 63 | |
GenBank mRNA | BC142415.1 | - | 3 - 4 | 2771 | 63 | |
BC151288.1 | - | 3 - 4 | 2518 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |