Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIGL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100311.2 | NP_001093781.1 | 1 - 2 | 250 | 108 | |
GenBank mRNA | BC149604.1 | - | 1 - 2 | 252 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |