Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RIPK4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100327.1 | NP_001093797.1 | 7 - 8 | 1199 | 74 | |
GenBank mRNA | BC148094.1 | - | 7 - 8 | 1199 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |