Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PJA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100340.1 | NP_001093810.1 | 7 - 8 | 1903 | 72 | |
GenBank mRNA | BC150108.1 | - | 7 - 8 | 1903 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |