Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP2S1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100366.1 | NP_001093836.1 | 7 - 8 | 1367 | 87 | |
GenBank mRNA | BC147910.1 | - | 7 - 8 | 1367 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |