Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OLFM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101879.2 | NP_001095349.1 | 1 - 2 | 263 | 112 | |
GenBank mRNA | BC149341.1 | - | 1 - 2 | 264 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |