Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102083.1 | NP_001095553.1 | 4 - 5 | 1459 | 113 | |
GenBank mRNA | BC149056.1 | - | 4 - 5 | 1459 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |