Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRPH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102378.1 | NP_001095848.1 | 8 - 9 | 1398 | 80 | |
GenBank mRNA | BC149862.1 | - | 8 - 9 | 1398 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |