Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C8H9orf72 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103088.1 | NP_001096558.1 | 5 - 6 | 777 | 79 | |
GenBank mRNA | BC150106.1 | - | 5 - 6 | 777 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |