Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C8H9orf72 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103088.1 | NP_001096558.1 | 6 - 7 | 890 | 116 | |
GenBank mRNA | BC150106.1 | - | 6 - 7 | 890 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |