Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GGH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105394.1 | NP_001098864.1 | 8 - 9 | 869 | 88 | |
GenBank mRNA | BC134544.1 | - | 8 - 9 | 869 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |