Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPRG1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076418.2 | NP_001069886.2 | 3 - 4 | 524 | 92 | |
GenBank mRNA | BC109547.1 | - | 3 - 4 | 427 | 92 | |
BT030599.2 | - | 3 - 4 | 523 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |