Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PARD6B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098104.1 | NP_001091573.1 | 1 - 2 | 254 | 56 | |
GenBank mRNA | BC140683.1 | - | 1 - 2 | 254 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |