Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EMC7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099143.2 | NP_001092613.1 | 3 - 4 | 509 | 124 | |
GenBank mRNA | BC134707.1 | - | 3 - 4 | 518 | 124 | |
BC142029.1 | - | 3 - 4 | 510 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |