Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NOX5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101137.1 | NP_001094607.1 | 8 - 9 | 1470 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Bos taurus Probe spans exons |