Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASB16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101188.1 | NP_001094658.1 | 1 - 2 | 378 | 93 | |
GenBank mRNA | BC147924.1 | - | 1 - 2 | 378 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |