Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC39A9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101191.1 | NP_001094661.1 | 6 - 7 | 1371 | 83 | |
GenBank mRNA | BC148060.1 | - | 6 - 7 | 1371 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |